Learn More
Description
Vivapure™ LentiSELECT 1000 is a pump driven kit for VSV-G pseudotyped lentivirus purification. The rapid 6 hour protocol results in a recovery of 4-5 x 109 infective particles per ml (total volume 2mL) from 1000mL cell culture supernatant.
This kit is to be operated by a laboratory pump and contains all necessary buffers and ultrafiltration devices for optimal convenience. The traditional time consuming ultracentrifugation method is replaced by this fast and simple Vivapure LentiSELECT 1000 kit.
Typically, for a normal yielding vector, a 1000mL cell culture purified using this method will yield up to 4-5 x 109 infective particles in 1 ml (total volume 2mL).
- Fast
- Non-toxic
- Better purity than pelleting by ultracentrifugation
- Convienient
- Cost effective
Specifications
Specifications
| Product Type | Fast Trap Adenovirus Purification and Concentration Kit |
| Sample Size | 1000 mL of Lentivirus supernatants |
| Purification Target | 40 to 1000 mL Cell Cultures |
| For Use With (Application) | VSV-G Pseudotyped Lentiviral Vectors |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.