Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivapure™ LentiSELECT 40 Purification and Concentration Kit

Catalog No. 14558563 Shop All Sartorius Products
Change view
Click to view available options
Sample Size:
4 x 40 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
14-558-563 4 x 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-563 Supplier Sartorius Supplier No. VSLVPQ040
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast purification of up to 8 x 108 viral particles

Vivapure LentiSELECT 40 is optimally suited for lentivirus purification from up to 40 ml cell culture and contains all components necessary for four purifications. Up to 8 x 108 viral particles are recovered in less than one hour. In contrast to traditional ultracentrifugation methods, virus purification with Vivapure LentiSELECT is fast and simple, without the requirement of expensive equipment like an ultracentrifuge. Additionally, this chromatographic procedure leads to pure virus samples in contrast to the crude ultracentrifuge pellet, for achieving higher reproducibility and increased gene transfer efficiency.

  • Fast
  • Non-toxic
  • Better purity than pelleting by ultracentrifugation
  • Convienient
  • Cost effective
TRUSTED_SUSTAINABILITY

Specifications

Sample Size 4 x 40 mL
Purification Target 40 to 1000 mL Cell Cultures
For Use With (Application) VSV-G Pseudotyped Lentiviral Vectors
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.