Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivapure™ miniprep A / G spin columns

Catalog No. p-4187766 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
16 Pack
2 Pack
48 Pack
Each
Type:
Miniprep Spin Column
Spin Column
Matrix:
Protein A Resin
Protein G Resin
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Matrix
14-558-538 48 Pack Spin Column Protein A Resin
14-558-539 16 Pack Spin Column Protein A Resin
14-558-543 48 Pack Spin Column Protein G Resin
14-558-540 Each Miniprep Spin Column Protein G Resin
14-558-544 16 Pack Spin Column Protein G Resin
14-558-545 2 Pack Miniprep Spin Column Protein A Resin
14-558-546 2 Pack Miniprep Spin Column Protein G Resin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-558-538 Supplier Sartorius Supplier No. VSARAMINIB

Centrifugal units with Protein A or Protein G resin for fast affinity purification of antibodies from small samples up to 0.65 ml.

Vivapure miniprep and spin columns are pre-packed centrifugal devices for purification of monoclonal and polyclonal antibodies from serum, ascites and cell culture supernatants. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed in parallel. Vivapure miniprep & spin columns can conveniently be used in a centrifuge with swing-out rotor for standard 2.2 ml tubes. Binding capacity is about 1 mg IgG per unit. The low hold-up volume ensures high solute recovery with minimal loss by non-specific adsorption. Vivapure miniprep and spin columns can be re-used three times and are available as complete kits including buffers or as bulk packs.They are an attractive alternative to lengthy chromatographic methods such as FPLC. For larger volumes, the Vivapure Maxiprep kit is also availablity.

  • Spin column based kit
  • Devices are ready to use
  • Rapid purification with high yields
  • Parallel processing
  • Cost effective through re-use option
  • Starter kits available

TRUSTED_SUSTAINABILITY

Specifications

Matrix Protein A Resin
Quantity 48 Pack
For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants
Type Spin Column
Sufficient For 3 Users
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.