Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivapure™ Q Mini H IEX Spin Columns

Catalog No. 14558553 Shop All Sartorius Products
Change view
Click to view available options
Capacity (Metric):
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-558-553 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-553 Supplier Sartorius Supplier No. VSIX01QH24
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use centrifugal devices based on Sartobind Membrane Adsorber technology as their chromatography matrix

Vivapure Mini Ion Exchange (IEX) spin columns are centrifugal devices, incorporating Sartobind Membrane Adsorber technology as their chromatography matrix. Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel. VIvapure Mini is available in Quaternary ammonium (Q) membrane chemistry.

Typical Applications

  • Sample preparation prior to 1-D or 2-D PAGE
  • Purification scouting of an unknown protein
  • Removal of endotoxins from monoclonal antibodies
  • Preparation of heme moiety from heme containing protein for functional analysis
  • Polishing of His-tagged proteins
  • General protein purification
  • Detergent removal from protein solutions
  • Purification of antibodies from serum, ascites or tissue culture supernatant
  • Establishing a purification/ pre-purification protocol for a given protein
  • HPLC/FPLC sample preparation
  • Purification of membrane-bound proteins
  • Fast centrifugal spin columns
  • Easy and ready-to-use
  • Binding capacity 1mg up to 4 mg protein
  • Ideal for parallel processing
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 240 μL
Capacity (Metric) 0.4 mL
Material Polypropylene
Pore Size 3 to 5 μm
Support Type Stabilized regenerated Cellulose
Quantity 24/Pk.
For Use With (Application) Sample fractionation, purification condition scouting, small scale purification
Type Spin Column
Sample Volume 24 x 0.5 mL
Ligand Type Quaternary Ammonium
Sufficient For Protein Purification
Column Type Ion Exchange Spin Column
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.