Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SASS6 Antibody [Alexa Fluor Plus 405], R&D Systems™
SDP

Catalog No. RD10016252
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10016252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10016252 Supplier R&D Systems Supplier No. NB10093342AFP405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems™. Rabbit Polyclonal Antibody

SASS6 Polyclonal antibody specifically detects SASS6 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen SASS6
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate Alexa Fluor Plus 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp761A078, FLJ22097, HsSAS-6, MGC119440, SAS-6, SAS6hsSAS-6, spindle assembly 6 homolog (C. elegans), spindle assembly abnormal protein 6 homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human spindle assembly 6 using the numbering given in entry NP_919268.1 (GeneID 163786).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 163786
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.