Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAT2 Monoclonal Antibody (OTI2B3)
GREENER_CHOICE

Catalog No. PIMA525940
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525940 100 μL
1 options

Catalog No. PIMA525940

Supplier: Invitrogen™ MA525940

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAT2 Monoclonal Antibody for Western Blot, IHC (P)

SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B3
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sat2
Gene Accession No. Q6P8J2, Q96F10
Gene Alias 2610016A03Rik; Diamine acetyltransferase 2; diamine N-acetyltransferase 2; EGK_08139; Polyamine N-acetyltransferase 2; Sat2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; spermidine/spermine N1-acetyltransferase family member 2; Ssat2; SSAT-2; thialysine N-epsilon-acetyltransferase
Gene Symbols Sat2
Host Species Mouse
Immunogen Full length human recombinant protein of SAT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112483, 69215
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.