Learn More
Abcam SATA (N-Succinimidyl-S-acetylthioacetate), Protein modification thiolating reagent, 250MG
Supplier:  Abcam AB145622250MG
Encompass_Preferred
MW 231.23 Da, Purity >94%. Primary amine-reactive protein modification thiolating reagent. Adds protected sulfhydryls to targeted proteins. Induces stable recombinant TNF-α trimer formation by formation of disulfide bridges between reactive thiol groups on adjacent TNF-α molecules, which show increased therapeutic efficacy in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5101
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.