Learn More
SATB2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301864AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Human special AT-rich sequence-binding protein-2 (SATB2) is a nuclear matrix/scaffold-associated region DNA-binding protein. Like its homolog SATB1, SATB2 selectively binds double-stranded, special AT-rich DNA sequences, but is expressed primarily in a subset of postmitotic, differentiating neurons in the neocortex. Mice deficient in SATB exhibit craniofacial abnormalities and defects in osteoblast differentiation and function. SATB2 also interacts with and enhances the activity of Runx2 and ATF4, two transcription factors that regulate osteoblast differentiation, indicating that SATB2 acts as a molecular node in a transcriptional network regulating skeletal development and osteoblast differentiation. Recent experiments have shown that SATB2 interacts with histone deacetylase 1 and metastasis-associated protein 2, two proteins that are involved in chromatin remodeling, suggesting that SATB2 may also be involved in mediating epigenetic influences during cortical development.Specifications
| SATB2 | |
| Polyclonal | |
| Unconjugated | |
| SATB2 | |
| FLJ21474, FLJ32076, KIAA1034, MGC119474, MGC119477, DNA-binding protein SATB2; SATB family member 2; special AT-rich sequence-binding protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23314 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9UPW6 | |
| SATB2 | |
| Between 250 and 300 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.