Learn More
AdipoGen Sauchinone (5 mg)
SDP
Supplier:  AdipoGen AGCN20032M005
Encompass_Preferred
Chemical. CAS: 177931-17-8. Formula: C20H20O6. MW: 356.4. Isolated from Saururus chinensis. Hepatoprotective antioxidant. Anti-inflammatory. Suppresses NF-kB p65 activity. Neuroprotective. Apoptosis inhibitor. Bone resorption/osteoclastogenesis inhibitor. Reduces ROS formation. Antiseptic. AMP-activated kinase (AMPK) activator. Akt phosphorylation inhibitor.
Catalog No. 50-205-3378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.