Learn More
QuidelOrtho™ Savanna™ HSV 1+2/VZV Assay 12KT
SDP
Catalog No. 23044186 Shop All QuidelOrtho Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
Catalog No. Form
23-044-186 Ready-to-Use (RTU)
1 options
This item is not returnable. View return policy
Catalog No. 23-044-186 Supplier QuidelOrtho™ Supplier No. 20445
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Savanna HSV 1+2/VZV Assay is an automated, rapid multianalyte real-time PCR test intended for use with the Savanna Platform and approved for the simultaneous qualitative detection and differentiation of herpes simplex virus type 1, herpes simplex virus type 2 and varicella-zoster virus.
  • The Savanna HSV 1+2/VZV Assay is a triplex assay that quickly distinguishes between three distinct herpes viruses: Herpes simplex virus 1, herpes simplex virus 2 and varicella-zoster, the virus that causes chicken pox and shingles infections
  • Why perform a triplex assay?
    • Visual differentiation of herpes lesions is not possible
    • Herpes lesions can present in atypical anatomic locations
    • Distinguish between HSV-1, HSV-2, and VZV in under 25 minutes withone patient sample
    • Timely diagnosis is highly effective in reducing onward transmission
    • Helps in patient counseling and treatment decisions
Specifications
CE Marker CE-IVD
Clia Complexity Moderate (Waiver Pending)
Detectable Analytes HSV
Form Ready-to-Use (RTU)