Learn More
QuidelOrtho™ Savanna™ Instrument, For Lease
SDP
Catalog No. 23044187 Shop All QuidelOrtho Products
Change view
Click to view available options
Product Type:
Savanna Molecular System
Catalog No. Product Type
23-044-187 Savanna Molecular System
1 options
This item is not returnable. View return policy
Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Catalog No. 23-044-187 Supplier QuidelOrtho™ Supplier No. 20382L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Savanna™ is a compact benchtop instrument that utilizes real-time PCR to deliver automated customized molecular results in minutes.
  • Superior performance and workflow
  • Utilizes real-time PCR technology
  • Simple sample-to-result workflow and minimal hands-on time
  • No upfront sample or reagent preparation
  • Results in 25 minutes or less
  • Customizable multiplex testing
  • Multiplex test panels, up to 12 targets per cartridge
  • Customizable assay panels to test for the most clinically relevant targets using the exclusive Test Select feature
  • Unlock masked test results within 48 hours without the need to collect a new sample or retest
  • Compact instrument design and simple workflow allows near-patient decentralized testing
  • Affordability
  • Multiple acquisition options
  • No service agreement required
  • Competitively priced panels and robust assay pipeline
Specifications
Product Type Savanna Molecular System