Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
GSA_VA
Catalog No. 20548137
Change view
Click to view available options
Type:
Acid Neutralization Cartridge
Ammonia Neutralizing Solution
Chemical Trap
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Mist Filter Replacement Cartridge
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
20-548-137 Condensation Flask
20-548-141 Chemical Trap
20-548-142 Chemical Trap Kit
20-548-144 Disposable Cartridge
20-548-143 Acid Neutralization Cartridge
20-548-145 Ammonia Neutralizing Solution
16-600-196 Heat Transfer Fluid
75-207-119 Mist Filter Replacement Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 20-548-137 Supplier Thermo Scientific™ Supplier No. GCF400
Add to Cart
Add to Cart
This item is not returnable. View return policy
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Glass condensation flask
Type Condensation Flask
For Use With (Equipment) SPD1030 and SPD2030 DNA SpeedVac™ Concentrator