Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors
Catalog No. p-4529001
Change view
Click to view available options
Holds:
2 Shallow-well Plates
36
48
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
20-548-128 2 Shallow-well Plates
20-548-104 36
20-548-105 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-548-128 Supplier Thermo Scientific™ Supplier No. RD2MP
Add to Cart
Edge
Add to Cart
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
TRUSTED_SUSTAINABILITY
Specifications
Description 2 shallow well plates
Type Concentrator Rotor
For Use With (Equipment) DNA130
Holds 2 Shallow-well Plates
Rotor System Traditional