Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD120 Vacuum Concentrator and Kits
Description
These modular vacuum concentrators provide flexibility for your benchtop space. They are ideal for evaporation of non-aggressive organics. Choose from one of 2 kits available: SPD120P1 SpeedVac kit for standard evaporation or SPD120 P2 SpeedVac™ kit for lyophilization. All SpeedVac kits contain all the necessary components under one model number.
Description
Space-saving design:
- Automatic bleeder valve for un-attended start/stop
- Digital display of parameters for easy, reproducible selection
- Three user defined programs that can be modified and saved
- Independent timers for both heat and run and audible alarm when run is completed
- PTFE-coated chamber for superior chemical resistance and easy cleaning
- Match up with an appropriate cold trap, vacuum pump or chemical trap for a complete concentration/drying system
Product Features:
- Basic molecular biology applications with increased microplate capacity
- Simple display and programming
- Six programs available
- Enhanced evaporation model, organic compatible
- Radiant glass cover to accelerate evaporation and reduce drying time
- Vacuum end point mode terminates run when preset vacuum level is reached
- Digital display of vacuum, temperature and time section
- Latched lid and lid lock indicator ensuring user safety during operation
SpeedVac SPD120 P1 Kit includes:
- SpeedVac concentrator
- Universal vacuum system (UVS450) including an oil-free vacuum pump and a -50 °C refrigerated vapor trap
- Glass condensation flask (GCF400)
- 40 x 1.5 mL rotor (RH40-11)
- Tubing package including 12’ of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
- CryoCool heat transfer fluid (1 L)
SpeedVac SPD120 P2 Kit includes:
- SpeedVac concentrator, basic molecular biology
- -105 ºC, ultra-low temperature refrigerated vapor trap
- VPL120 high vacuum pump
- 40 x 1.5 mL rotor (RH40-11)
- Universal tubing package (TFK100)
- Digital vacuum gauge
- Chemical trap kit for use with disposable cartridge
- Disposable cartridge with color indicator for trapping acids and water vapor
- Glass condensation flask
- Vacuum pump lubricating oil
- CryoCool™ heat transfer fluid (1 L)
Solvent and solvent combinations
- Acetonitrile
- Ethanol
- Isopropanol
- Methanol
- Water
- Acetonitrile and water
- Ethanol and water
- Methanol and water
- Ammonium hydroxide
Typical applications:
- Drying biomolecules in HPLC fractions
- Evaporating purification solvents
- Drying extracts of natural products in drug assays
- Lyophillization
Specifications
Specifications
| Includes | Kit for Standard Evaporation |
| Certifications | UL Listed to US and Canadian requirements. CE marked |
| Description | SpeedVac SPD120 Vacuum Concentrator Kit |
| Electrical Requirements | 230 V 50 Hz |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.