Learn More
Ansell™ Sawyer-Tower™ 66-677 Series Coveralls
Catalog No. 19010049 Shop All Ansell Products
Change view
Click to view available options
:
XX-large
Catalog No.
19-010-049 XX-large
1 options
This item is not returnable. View return policy
Catalog No. 19-010-049 Supplier Ansell™ Supplier No. 666772XL
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Full-coverage chemical protection with added safety of fire resistance
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistance
  • HRC rating: 2
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material CPC Nomex™ Tri-laminate
Color Blue
Resistance Flame, Chemical
Arc Flash Rating 2