Learn More
TARGETMOL CHEMICALS INC SB 218795 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T23316
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB 218795 is an effective and selective antagonist of NK3 with a Ki 13 nM for hNK3 which is about 90-fold and 7000-fold selectivity over hNK2 and hNK1. purity: 98%
Catalog No. 50-258-0882
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.