Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 222200
GREENER_CHOICE

Catalog No. p-4785525
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-931-0 10 mg
13-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-931-0 Supplier Tocris Bioscience™ Supplier No. 1393/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective non-peptide NK3 antagonist. Brain penetrant

Potent and selective non-peptide NK3 receptor antagonist (Ki values are 4.4, > 100,000 and 250 nM for human NK3, NK1 and NK2 receptors respectively). Antihypertensive in vivo. Brain penetrant.

Chemical Identifiers

CAS 174635-69-9
Molecular Formula C26H24N2O

Specifications

Quantity 10 mg
Formula Weight 380.48
Purity >98%
Molecular Weight (g/mol) 380.491
Chemical Name or Material SB 222200
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.