Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 269970 hydrochloride
GREENER_CHOICE

Catalog No. 161210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-121-0 10 mg
16-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-121-0 Supplier Tocris Bioscience™ Supplier No. 1612/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT7 antagonist; brain penetrant

Potent and selective 5-HT7 receptor antagonist (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B and < 6.0 for 5-HT1A, 5-HT1D, 5-HT1E, 5-HT1F, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT4 and 5-HT6 receptors respectively). Brain penetrant in vivo.

Chemical Identifiers

CAS 261901-57-9
Molecular Formula C18H29ClN2O3S

Specifications

Quantity 10 mg
Formula Weight 388.95
Purity >98%
Molecular Weight (g/mol) 388.951
Chemical Name or Material SB 269970 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.