Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 328437
GREENER_CHOICE

Catalog No. 365010R
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-501-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 36-501-0R Supplier Tocris Bioscience™ Supplier No. 3650/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CCR3 antagonist

SB 328437 is a potent and selective CCR3 antagonist (IC50 = 4 nM). Displays > 2500-fold selectivity over C5aR, LTD4, CCR7, CXCR1 and CXCR2 receptors. Inhibits eotaxin-, eotaxin-2- and MCP-4-induced Ca2+ mobilization (IC50 values are 38, 35 and 20 nM respectively) and inhibits eotaxin-, eotaxin-2- and MCP-4-induced eosinophil chemotaxis with similar potencies.

Specifications

Quantity 10 mg
Product Type SB 328437
Purity >99%
Molecular Weight (g/mol) 378.38
Inhibitors SB 328437

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.