Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 334867
GREENER_CHOICESDP

Catalog No. 196010
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-601-0 10 mg
19-605-0 50 mg
19-601-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-601-0 Supplier Tocris Bioscience™ Supplier No. 1960/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective non-peptide OX1 antagonist

SB 334867 is a selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.

Specifications

Chemical Name or Material N-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea
CAS 792173-99-0
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 100mM with gentle warming in DMSO
Molecular Formula C17H13N5O2
Formula Weight Observed MW: 319.32

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.