Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 431542
GSA_VAGREENER_CHOICE

Catalog No. 161410
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective inhibitor of TGF-βRI, ALK4 and ALK7

SB 431542 is a potent and selective inhibitor of the transforming growth factor-β (TGF-β) type I receptor/ALK5 (IC50 = 94 nM), and its relatives ALK4 and ALK7. Suppresses TGF-β-induced proliferation of human osteosarcoma cells. Replaces SOX2 in reprogramming of fibroblasts into iPSCs. Stimulates proliferation, differentiation and sheet formation of ESC-derived endothelial cells. Inhibits TGF-β-induced EMT, migration, invasion and VEGF secretion in several human cancer cell lines.

SB 431542 synthesized to cGMP guidelines also available.

For more information about how SB 431542 may be used, see our protocols: 3D Culture of Lung Alveolar Cells, Differentiation of Hematopoietic Progenitors from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Generating Vascular Organoids, Accelerated Induction of Cortical Neurons from hiPSCs .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material SB 431542
CAS 301836-41-9
Synonym 4-4-2h-1,3-benzodioxol-5-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide
Quantity 10 mg
Molecular Formula C22H16N4O3
Formula Weight 384.39
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.