Learn More
Medchemexpress LLC HY-13521 50mg , SB-505124 CAS:694433-59-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1352150MG
Encompass_Preferred
Medchemexpress, HY-13521 50mg SB-505124 CAS:694433-59-5 Formula:C20H21N3O2 IC50: 129 nM (ALK4), 47 nM (ALK5) Purity:98% SB-505124 is a selective inhibitor of  TGFβR with  IC 50  of 129 nM and 47 nM for  ALK4 , ALK5 , respectively, and it does not inhibits ALK1, 2, 3, or 6 but ALK7. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3770
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.