Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 505124
GREENER_CHOICE

Catalog No. p-4784504
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-631-0 10 mg
32-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-631-0 Supplier Tocris Bioscience™ Supplier No. 3263/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of TGF-βRI, ALK4 and ALK7

Selective inhibitor of transforming growth factor-β type I receptor (ALK5), ALK4 and ALK7 (IC50 values are 47 and 129 nM for ALK5 and ALK4 respectively). Selectively inhibits signaling from TGF-β and activin; does not inhibit other ALK family members.

Chemical Identifiers

CAS 694433-59-5
Molecular Formula C20H21N3O2

Specifications

Quantity 10 mg
Formula Weight 335.4
Purity >98%
Molecular Weight (g/mol) 335.407
Chemical Name or Material SB 505124
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.