Learn More
AdipoGen SB216763 (25 mg)
SDP
Supplier:  AdipoGen AGCR13659M025
Encompass_Preferred
Chemical. CAS: 280744-09-4. Formula: C19H12Cl2N2O2. MW: 371.2. Synthetic. Potent and selective cell permeable, ATP-competitive inhibitor of GSK3alpha with an IC50 value of 34nM. DYRK1A inhibitor (IC50=0.8µM). Stimulates glycogen synthesis in liver cells and induces beta-catenin-dependent gene transcription. Acts as neuroprotectant and prevents neuronal cell death induced by PI3-kinase pathway. Anticancer agent with antiproliferative activity. Reduces pulmonary inflammation and fibrosis in a mouse model. Shown to delay preconditioning, reduces infarct size and prevents cardiac ischemia. Promotes pluripotency in mouse embryonic stem cells. Glycogen synthase kinase 3 (GSK3) is a serine/threonine protein kinase that is inhibited by an assortment of extracellular stimuli such as insulin, growth factors, cell specification factors and cell adhesion. Its activity regulates many cell functions including the control of cell division, apoptosis and inflammation.
Catalog No. 50-205-4040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.