Learn More
Selleck Chemical LLC SB431542 | 99.99% | 301836-41-9 | 1g
SDP
Supplier:  Selleck Chemical LLC S10671G
Encompass_Preferred
SB431542 is a potent and selective inhibitor of ALK5 with IC50 of 94 nM in a cell-free assay, 100-fold more selective for ALK5 than p38 MAPK and other kinases.
Catalog No. NC2314879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.