Learn More
Tocris Bioscience™ sBADA
Description
Specifications
Specifications
| Chemical Name or Material | (R)-2-Amino-3-(3-(5,5-difluoro-7,9-dimethyl-2-sulfo-5H-4λ4,5λ4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanamido)propanoic acid |
| CAS | 2376838-11-6 |
| Quantity | 1 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C17H21BF2N4O6S |
| Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.