Learn More
AdipoGen SBD-F
SDP
Supplier:  AdipoGen CDXF0002M025
Encompass_Preferred
Chemical. CAS 84806-27-9. Formula C6H6FN3O4S. MW 235.19. Synthetic. Fluorescent probe for thiols widely used in pre-column labeling of biological thiols for HPLC.
Catalog No. 50-256-0954
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.