Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SBD-F, Fluorescent Thiol Indicator, 235.2 Da, >98% Purity, 250 mg

Catalog No. 502705227
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5227 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5227 Supplier Abcam Supplier No. AB145405250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorescent thiol indicator that reacts with sulfhydryl groups to produce highly fluorescent compounds for low detection limits.

  • Molecular weight: 235.2 Da
  • Purity: >98%
  • Application: Fluorescent thiol indicator; reacts with sulfhydryl groups to produce highly fluorescent compounds
  • Performance: Attains low detection limits

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Sulfhydryl Groups
Substrate Properties Chemical Substrate
Substrate Sulfhydryl groups
Shipping Condition Ambient
Detection Method Fluorescence
Buffer 100% SBD-F
Content And Storage Store at -20°C.
Form Powder
Reactivity Sulfhydryl groups
Formulation 100% SBD-F
Quantity 250 mg
For Use With (Application) Thiol detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.