Learn More
Abcam SBD-F, Fluorescent Thiol Indicator, >98% Purity, 25 mg
Description
- Molecular Weight: 235.2 Da
- Purity: >98%
- Description: Fluorescent thiol indicator that reacts with sulfhydryl groups to produce highly fluorescent compounds.
- Performance: Attains low detection limits.
- Storage: Store at -20°C.
- Composition: 100% SBD-F
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% SBD-F |
| Target Molecule | Sulfhydryl Groups |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Reactivity | Sulfhydryl groups |
| Shipping Condition | Ambient |
| Substrate | Sulfhydryl groups |
| Detection Method | Fluorescence |
| Formulation | 100% SBD-F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.