Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SBFI AM, Cell-Permeable, Fluorescent Na+ Indicator, 1 mg

Catalog No. 502583313
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3313 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3313 Supplier Abcam Supplier No. AB1428001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable fluorescent probe for sodium ion detection.

  • Molecular weight: 1127.1 Da
  • Purity: >95%
  • Indicator type: Cell-permeable, fluorescent Na+ indicator
  • Functionality: Binds to Na+ ions (Kd = 7.4 mM); fluorescence intensity increases upon Na+ binding

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Sodium Ions
Substrate Properties Chemical Substrate
Substrate Na+
Shipping Condition Ambient
Detection Method Fluorescence
Detection Location In-Cell Detection
Buffer 100% bis(acetyloxymethyl) 4-[6-[13-[2-[2,4-bis(acetyloxymethoxycarbonyl)phenyl]-5-methoxy-1-benzofuran-6-yl]-1,4,10-trioxa-7,13-diazacyclopentadec-7-yl]-5-methoxy-1-benzofuran-2-yl]benzene-1,3-dicarboxylate
Content And Storage Store at -20°C.
Quantity 1 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.