Learn More
Selleck Chemical LLC SBI-0640756
SDP
Supplier:  Selleck Chemical LLC S81811G
Encompass_Preferred
SBI-0640756(SBI-756) is a first-in-class inhibitor that targets eIF4G1 and disrupts the eIF4F complex It can also suppress AKT and NF- B signaling
Catalog No. 50-000-01904
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.