Learn More
PIP™ SBP Bouffant 9 gsm
Catalog No. 17377591 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Green
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-377-591 Red
17-377-590 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-377-591 Supplier PIP™ Supplier No. UB241000R
Add to Cart
Add to Cart
Polypropylene, disposable, and 24 in. SBP Bouffant
  • Polypropylene disposable clothing provides affordable, lightweight protection against non-hazardous dry particulates like dirt.
  • Good for use in pharmaceuticals, medical, law enforcement and laboratory.
Specifications
Product Type Clip Cap
Size 24 in.
Material Spunbond Polypropylene
Color Red
Disposable Single-use
Latex Free Yes
Sterility Non-sterile