Learn More
Mascon Medical Mascon Medical™ SBS Storage Tube, 4.0ml, 50EA/Bag, 25 Bags/Case
SDP
Supplier:  Mascon Medical FSSBSTUBE4
Encompass_Preferred
Our Mascon Medical™ SBS Storage Tubes are made from medical grade polypropylene and are resistant to high temperature and high pressure and can be frozen and thawed repeatedly. Our storage tubes come in a variety of different colored caps to easily identify samples with writing area to label samples. Features:•Meets ISO13485 quality management system standard•Can maintain -60Kpa negative pressure test without leakage•2D/1D bar code•External thread•Round bottom•Self-stand•RNase/DNase free•Non-pyrogenic•Endotoxin-freeSpecifications:•Volume: 4.0ml•Qty. / Case: 1,250•Material: Polypropylene•Sterile: No•Temperature Range: -196℃~120℃•Packaging Format: 50 EA/Bag, 25 Bags/Case
Catalog No. NC2458348
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.