Learn More
Enzo Life Sciences SC-514 (50mg). CAS: 354812-17-2
SDP
Supplier:  Enzo Life Sciences BMLEI3430050
Encompass_Preferred
Selective, reversible and ATP-competitive inhibitor of IKK2 (IC50=11.2µM). Does not inhibit other IKK isoforms or other serine/threonine or tyrosine kinases. Inhibits transcription of NF-κB dependent genes in IL-1β-induced rheumatoid arthritis-derived synovial fibroblasts in a dose-dependent fashion. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.