Learn More
Sunrise Science Products SC-His-Leu-Ura Powder, 30 grams
Supplier:  Sunrise Science Products 1327030
Encompass
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media. Mix with stirring for 10-15 minutes, and filter sterilize or autoclave at 121°C for 15 minutes. To prepare plates, autoclave agar separately and add to sterile medium, or add agar to liquid medium and adjust to pH 5.8-6.0 before autoclaving. Recipe in mg/L: Adenine hemisulfate (21 mg/L), L-Alanine (85.6 mg/L), L-Arginine (85.6 mg/L), L-Asparagine monohydrate (85.6 mg/L), L-Aspartic acid (85.6 mg/L), L-Cysteine hydrochloride monohydrate (85.6 mg/L), L-Glutamic acid (85.6 mg/L), L-Glutamine (85.6 mg/L), L-Glycine (85.6 mg/L), Inositol (in AA/Supplement Mix) (85.6 mg/L), L-Isoleucine (85.6 mg/L), L-Lysine hydrochloride (85.6 mg/L), L-Methionine (85.6 mg/L), 4-Aminobenzoic acid (PABA) (In AA/Supplement Mix) (8.6 mg/L), L-Phenylalanine (85.6 mg/L), L-Proline (85.6 mg/L), L-Serine (85.6 mg/L), L-Threonine (85.6 mg/L), L-Tryptophan (85.6 mg/L), L-Tyrosine (85.6 mg/L), L-Valine (85.6 mg/L).
Catalog No. NC1577490
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.