Learn More
Interscience Scan 50
SDP
Catalog No. 16948889 Shop All Interscience Products
Change view
Click to view available options
Includes:
HandPad, Headphones
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
16-948-889 -
16-949-092 HandPad, Headphones
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-948-889 Supplier Interscience Supplier No. 435050
Only null left
Add to Cart
Add to Cart
The Scan 50 is a manual colony counter with improved ergonomics.
Scan 50 is a manual colony counter with improved ergonomics that help prevent Musculoskeletal Disorders. It offers contrasting illumination of colonies while protecting the user's eyes. Traceability is ensured by a USB port.
  • Counting on: 55 to 90 mm dia. Petri dishes, Petrifilm™, MC-media Pads™, Compact Dry™, EasyPlate™, filtration membranes
  • Dark Field 2: Indirect LED lighting
  • Data export via USB, LIMS, under CSV format (Excel™) or text format (Word™)
The Scan 50 Pro includes headphones and an ergonomic arm rest.
Specifications
Product Type Darkfield Colony Counter
Display Type LED
Voltage 100/240 V