Learn More
Description
- EasySpiral and EasySpiral Dilute circle counting grids
- Diameter: 90 mm
- 1 transparent and 1 white diffusing background for Scan 50 and Scan 50 Pro
Specifications
Specifications
| Product Type | Counting Grids |
| Display Type | None |
| Color | Transparent and White |
| Product Line | Easy Spiral |
| Quantity | 2/Pk. |
| Diameter (Metric) | 90 mm |
| For Use With (Equipment) | Scan 50 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
