Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) bromide, ultra dry, 99.99% (metals basis)

Catalog No. p-7052327
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4719903 1 g
AA4719906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4719903 Supplier Thermo Scientific Chemicals Supplier No. 04719903
Only null left
Add to Cart

CAS: 13465-59-3 | Br3Sc | 284.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-59-3
Molecular Formula Br3Sc
Molecular Weight (g/mol) 284.67
MDL Number MFCD00016324
InChI Key APPHYFNIXVIIJR-UHFFFAOYSA-K
Synonym scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis
PubChem CID 83495
IUPAC Name scandium(3+) tribromide
SMILES [Sc+3].[Br-].[Br-].[Br-]

Specifications

Physical Form Powder
Quantity 1 g
Formula Weight 284.67
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Scandium(III) bromide, Ultra dry
EINECSNumber 236-699-6
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.