Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermco™ ScanTemp 410 Handheld Infrared Thermometer

Catalog No. 04500021
Change view
Click to view available options
Max. Temperature (Metric):
500°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. Temperature (Metric)
04-500-021 500°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-500-021 Supplier Thermco™ Supplier No. ACCD0503IR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ScanTemp 410 uses infrared technology to measure the temperature of materials up close, or from a distance.

You simple aim, pull the trigger, and read the temperature on a large, backlight LCD display. Lightweight, compact, and easy-to-use, the ScanTemp 410 can safety measure hot, hazardous, or hard-to-reach materials without contaminating or damaging the material's surface. For safe and accurate temperature measurement, reach for a ScanTemp 410.

APPLICATIONS:

  • HVAC
  • Electrical Maintenance
  • Road Construction
  • Food Safety (HACCP)
  • Commercial Printing
  • Plastics Molding
  • Car Maintenance
  • Detect Process Maintenance Problems
  • Pouch and 2 1.5 V batteries included

Specifications

Product Type IR Thermometer
Max. Temperature (English) 932°F
Max. Temperature (Metric) 500°C
Min. Temperature (English) -27°F
Min. Temperature (Metric) -33°C
Calibrated No
Accuracy ±2% or 2°C
Display Digital LCD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.