Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCARA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110903 Supplier Invitrogen™ Supplier No. PA5110903
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDDLKALTRN VNRLNESFRD LQLRLLQAPL QADLTEQVWK VQDALQNQSD SLLALAGAVQ RLE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCARA5
Gene Accession No. Q6ZMJ2
Gene Alias 4932433F15Rik; 4933425F03Rik; AV278087; NET33; SCARA5; Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); scavenger receptor hlg; Tesr; testis expressed scavenger receptor; UNQ2938/PRO28700
Gene Symbols SCARA5
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SCARA5. Recombinant protein control fragment (Product #RP-93888).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.