Learn More
Invitrogen™ SCARF1 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Specifications
Specifications
| Antigen | SCARF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Scarf1 |
| Gene Accession No. | Q14162 |
| Gene Alias | AA986099; acetyl LDL receptor; endothelial cells scavenger receptor; KIAA0149; MGC47738; mKIAA0149; SCARF1; Scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; SREC; SREC1; SREC-I |
| Gene Symbols | Scarf1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.