Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCC-112 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501556873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-6873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-6873 Supplier Bethyl Laboratories, Inc Supplier No. A300089A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

PDS5A belongs to the PDS5 family. It contains 1 HEAT repeat and 3 LRR (leucine-rich) repeats. PDS5A may contribute to regulation of sister chromatid cohesion during mitosis.

Specifications

Antigen SCC-112
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PDS5A
Gene Accession No. Q29RF7
Gene Alias cell proliferation-inducing gene 54 protein, KIAA0648, PDS5, regulator of cohesion maintece, homolog A, PDS5, regulator of cohesion maintece, homolog A (S. cerevisiae), PIG54, SCC112, SCC-112, sister chromatid cohesion protein 112
Gene Symbols PDS5A
Host Species Rabbit
Immunogen Between 1250 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23244
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.