Learn More
TARGETMOL CHEMICALS INC SCD1 INHIBITOR-3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T38683
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCD1 inhibitor-3 (ML-270) is a highly effective and orally available compound that inhibits SCD1 with remarkable safety. SCD1 inhibitor-3 demonstrates strong potential for research in metabolic diseases including obesity type II diabetes and dyslipidemia as well as various skin conditions like acne and cancer. purity: 98%
Catalog No. 50-258-0092
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.