Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech SCF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P71BT50
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P71BT50 50 μg
500P71BT25 25 μg
50-271-8457 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P71BT50 Supplier PeproTech Supplier No. 500P71BT50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine SCF. Anti-Murine SCF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mSCF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine SCF (500-P71) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mSCF. Western Blot: To detect mSCF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mSCF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P71BT-1MG will be provided as 2 x 500 μg

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Kitl
Gene Accession No. P20826
Gene Alias 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Gene Symbols Kitl
Host Species Rabbit
Immunogen E.coli-derived, 18.3 kDa Recombinant Murine SCF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17311
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.