Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552357
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA552357 100 μL
1 options

Catalog No. PIPA552357

Supplier: Invitrogen™ PA552357

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDGTPLTAED IVHKIAARIY EENDRAVFDK IVSKLLNLGL ITESQAHTLE DEVAEVLQKL ISKEANNYEE DPNKPTSWTE NQAGKIPEKV TPMAAIQDGL AKGE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Granins may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCG3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCG3
Gene Accession No. Q8WXD2
Gene Alias 1B1075; AI385542; Chgd; Scg3; Secretogranin III; secretogranin-3; SgIII; UNQ2502/PRO5990
Gene Symbols SCG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCG3. Recombinant protein control fragment (Product #RP-88664).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.