Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 51344

Catalog No. 528050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-805-0 50 mg
52-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-805-0 Supplier Tocris Bioscience™ Supplier No. 5280/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent MTH1 inhibitor

SCH 51344 is a potent MTH1 inhibitor (Kd = 49 nM). Inhibits Ras-induced malignant transformation and increases α-actin promoter-driven CAT activity in Ras-transformed cells. Has no effect on Ras-induced ERK and JNK activation. Inhibits Ras-induced membrane ruffling in REF-52 fibroblasts and blocks anchorage-independent growth of Ras-transformed tumor cell lines. Also induces DNA damage in SW480 colon cancer cells.

Specifications

Purity >99%
Quantity 50 mg
Product Type SCH 51344
Molecular Weight (g/mol) 316.35
Inhibitors SCH 51344

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.