Learn More
Abcam SCH-58261, A2A antagonist, 5MG
Supplier:  Abcam AB1204395MG
Encompass_Preferred
MW 345.4 Da, Purity >99%. Potent, highly selective A2A receptor antagonist. (Ki values are 1-2, 289 and >10000 nM at A2A, A1 and A3 receptors, respectively). Displays therapeutic effects in animal Parkinson's models and elicits locomotor sensitization *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7587
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.