Learn More
Cayman Chemical SCH 772984, 942183-80-4, 1 mg
SDP
Supplier:  Cayman Chemical 191661
Encompass_Preferred
A potent, selective inhibitor of ERK1 and ERK2 (IC50s = 4 and 1 nM, respectively); has nanomolar cytotoxicity in tumor cells with mutations in BRAF, NRAS, or KRAS; effective in vivo, inducing regression of xenograft tumors in mice
Catalog No. NC1529178
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.