Learn More
TARGETMOL CHEMICALS INC SCH-900271 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34573
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH-900271 is a potent nicotinic acid receptor for the treatment of dyslipidaemia and is commonly used in the study of mixed hyperlipidaemia and primary hypercholesterolaemia. purity: 98%
Catalog No. 50-258-0256
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.