Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ Schaedler K Agar (Sheep Blood 5%)
SDP

Catalog No. 21403470 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-470 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-470 Supplier Liofilchem™ Supplier No. 11065
Add to Cart
Add to Cart

A medium for anaerobic bacteria isolation, effective for isolating bacteria

  • An enriched, non-selective medium used for the isolation and cultivation of fastidious anaerobic and aerobic bacteria from clinical specimens.
  • It is particularly effective for isolating bacteria that are difficult to grow, such as Bacteroides, Prevotella, and Porphyromonas spp.
  • It is used with various clinical specimens, including genital samples, upper respiratory swabs, and gastrointestinal tract samples to identify potential pathogens.

Specifications

Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Anaerobes
Test Type Microbial Cultivation
Quantity 20 Plates
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.